Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNPO3 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB358310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358310 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358310 Supplier Novus Biologicals Supplier No. NBP176374PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TNPO3 Polyclonal antibody specifically detects TNPO3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen TNPO3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias imp12, MTR10A, transportin 3, transportin-3
Host Species Rabbit
Immunogen Antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human TNPO3. The immunogen is located within amino acids 220 - 270 of TNPO3. Amino Acid Squence: EVLQQDKSTSSNLHEAASD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23534
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.