Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOB1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP180225 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP180225 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP180225 Supplier Novus Biologicals Supplier No. NBP180225
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TOB1 Polyclonal specifically detects TOB1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen TOB1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_033453
Gene Alias APRO6, MGC104792, PIG49, protein Tob1, TOBMGC34446, Transducer of erbB-2 1, transducer of ERBB2, 1, TROB, TROB1
Gene Symbols TOB1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of mouse TOB1. Peptide sequence QPLTFTTATFAATKFGSTKMKNSGRSSKVARTSPINLGLTVNVNDLLKQK.
Molecular Weight of Antigen 40 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10140
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.