Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOE1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290719 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290719 Supplier Novus Biologicals Supplier No. NBP204022APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TOE1 Polyclonal antibody specifically detects TOE1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen TOE1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias FLJ13949, target of EGR1 protein 1, target of EGR1, member 1 (nuclear)
Host Species Rabbit
Immunogen Maps to a region between residue 460 and 510 of human Target of EGR1, Member 1 using the numbering given in entry NP_079353.3 (GeneID 114034).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114034
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.