Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tollip Antibody (SB40a), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP128621C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128621C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128621C Supplier Novus Biologicals Supplier No. NBP128621C

Mouse Monoclonal Antibody

Tollip Monoclonal specifically detects Tollip in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen Tollip
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone SB40a
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias adapter protein, FLJ33531, IL-1RAcPIP, toll interacting protein, toll-interacting protein
Gene Symbols TOLLIP
Host Species Mouse
Immunogen Recombinant Tollip
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54472.0
Test Specificity Human Tollip; Does not react to TLR2, TLR4, TLR6, TLR7, TLR8 and TLR10 recombinant proteins
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.