Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tollip Antibody (SB40a), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB284592 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB284592 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB284592 Supplier Novus Biologicals Supplier No. NBP128621MFV500

Mouse Monoclonal Antibody

Tollip Monoclonal antibody specifically detects Tollip in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Tollip
Applications Flow Cytometry
Classification Monoclonal
Clone SB40a
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias adapter protein, FLJ33531, IL-1RAcPIP, toll interacting protein, toll-interacting protein
Host Species Mouse
Immunogen Recombinant Tollip
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54472
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.