Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOM1L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167201 100 μg
NB167202 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167201 Supplier Novus Biologicals Supplier No. NBP316053100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TOM1L2 Polyclonal antibody specifically detects TOM1L2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen TOM1L2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias FLJ21501, FLJ32746, target of myb1 (chicken) homolog-like 1, target of myb1-like 2 (chicken), Target of Myb-like protein 2, TOM1-like protein 2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 310-450 of human TOM1L2 (NP_001076437.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 146691
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.