Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOM70 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB11058347
Description
TOM70 Polyclonal specifically detects TOM70 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.Specifications
TOM70 | |
Polyclonal | |
Western Blot 1:500, Simple Western 1:50, Immunocytochemistry/Immunofluorescence 1:100 | |
O94826 | |
TOMM70 | |
A synthetic peptide made to a region within residues 1-100 of the human TOM70 protein. [Swiss-Prot# O94826] | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS with No Preservative | |
FLJ90470, KIAA0719, mitochondrial import receptor subunit TOM70, Mitochondrial precursor proteins import receptor, TOM70, Translocase of outer membrane 70 kDa subunit, translocase of outer mitochondrial membrane 70 (yeast) homolog A, translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae), translocase of outer mitochondrial membrane 70 homolog A (yeast) | |
Rabbit | |
67 kDa | |
0.1 mL | |
Apoptosis, Cellular Markers, Mitochondrial Markers, Signal Transduction | |
9868 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction