Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOMM20 Antibody (1B2), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB163075 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163075 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163075 Supplier Novus Biologicals Supplier No. NBP260248G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TOMM20 Monoclonal antibody specifically detects TOMM20 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen TOMM20
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 1B2
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias KIAA0016Outer mitochondrial membrane receptor Tom20, MAS20, MGC117367, Mitochondrial 20 kDa outer membrane protein, mitochondrial import receptor subunit TOM20 homolog, MOM19, TOM20, translocase of outer mitochondrial membrane 20 homolog (yeast), translocase of outer mitochondrial membrane 20 homolog type II
Host Species Mouse
Immunogen Recombinant human Tomm20 (25-145aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9804
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.