Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOMM40 Antibody, Novus Biologicals™
SDP

Catalog No. NB186575 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186575 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB186575 Supplier Novus Biologicals Supplier No. NBP312425
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TOMM40 Polyclonal antibody specifically detects TOMM40 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen TOMM40
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias C19orf1mitochondrial import receptor subunit TOM40 homolog, D19S1177E, mitochondrial outer membrane protein, PER-EC1, PEREC1Translocase of outer membrane 40 kDa subunit homolog, Protein Haymaker, TOM40p38.5, translocase of outer mitochondrial membrane 40 homolog (yeast)
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 300-350 on human Mitochondrial import receptor subunit TOM40 homolog.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10452
Target Species Human
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.