Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOMM40L Rabbit anti-Human, Alexa Fluor 405, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB045088 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045088 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045088 Supplier Novus Biologicals Supplier No. NBP297573AF405

Rabbit Polyclonal Antibody

TOMM40L Polyclonal specifically detects TOMM40L in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TOMM40L
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias FLJ12770, mitochondrial import receptor subunit TOM40B, Protein TOMM40-like, TOMM40BRP11-297K8.10, translocase of outer mitochondrial membrane 40 homolog (yeast)-like, translocase of outer mitochondrial membrane 40 homolog-like, translocase of outer mitochondrial membrane 40 homolog-like (yeast)
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TOMM40L fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84134
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.