Learn More
Description
Specifications
Specifications
| Antigen | TOMM7 |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:150 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | homolog of Tom7, mitochondrial import receptor subunit TOM7 homolog, TOM7, TOMM07,6.2 kd protein, Translocase of outer membrane 7 kDa subunit homolog, translocase of outer mitochondrial membrane 7 homolog (yeast) |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide taken within amino acid region 1-50 on human Mitochondrial import receptor subunit TOM7 homolog protein. |
| Purification Method | Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
