Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOMM7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$515.50
Specifications
| Antigen | TOMM7 |
|---|---|
| Dilution | Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:150 |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TOMM7 Polyclonal antibody specifically detects TOMM7 in Human, Mouse, Rat, Porcine, Bovine, Primate samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| TOMM7 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Endocrinology | |
| Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide | |
| 54543 | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:150 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Primate | |
| homolog of Tom7, mitochondrial import receptor subunit TOM7 homolog, TOM7, TOMM07,6.2 kd protein, Translocase of outer membrane 7 kDa subunit homolog, translocase of outer mitochondrial membrane 7 homolog (yeast) | |
| Synthetic peptide taken within amino acid region 1-50 on human Mitochondrial import receptor subunit TOM7 homolog protein. | |
| Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title