Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOP2A Antibody (TOP2A/4397R), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB384651 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384651 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384651 Supplier Novus Biologicals Supplier No. NBP308756AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TOP2A Monoclonal antibody specifically detects TOP2A in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen TOP2A
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TOP2A/4397R
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias DNA topoisomerase (ATP-hydrolyzing), DNA topoisomerase 2-alpha, DNA topoisomerase II, 170 kD, DNA topoisomerase II, alpha isozyme, EC 5.99.1.3, TOP2DNA gyrase, topoisomerase (DNA) II alpha (170kD), topoisomerase (DNA) II alpha 170kDa, TP2A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to TOP2A residues within aa1431-1531 of TOP2A (Uniprot: P11388 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7153
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.