Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOP2B Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB047753 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047753 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047753 Supplier Novus Biologicals Supplier No. NBP297789IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TOP2B Polyclonal specifically detects TOP2B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TOP2B
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias antigen MLAA-44, DNA topoisomerase 2-beta, DNA topoisomerase II beta, DNA topoisomerase II, 180 kD, DNA topoisomerase II, beta isozyme, EC 5.99.1.3, top2beta, TOPIIB, topo II beta, topoisomerase (DNA) II beta (180kD), topoisomerase (DNA) II beta 180kDa, topoisomerase II beta, topoisomerase IIb, U937 associated antigen
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TOP2B fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 7155
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.