Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOPAZ1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB183979 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183979 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183979 Supplier Novus Biologicals Supplier No. NBP282040AF647
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TOPAZ1 Polyclonal antibody specifically detects TOPAZ1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TOPAZ1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias Chromosome 3 Open Reading Frame 77, Testis And Ovary Specific PAZ Domain Containing 1, Testis And Ovary-Specific PAZ Domain Gene 1, Testis- And Ovary-Specific PAZ Domain-Containing Protein 1, Topaz 1, TOPAZ1
Host Species Rabbit
Immunogen TOPAZ1 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human TOPAZ1. The immunogen is located within amino acids 1330 - 1380 of TOPAZ1. Amino Acid Squence: EFAETVSKDPQNSKVDK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 375337
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.