Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TopBP1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB292484 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292484 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292484 Supplier Novus Biologicals Supplier No. NB100217PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TopBP1 Polyclonal antibody specifically detects TopBP1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen TopBP1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias DNA topoisomerase II-beta-binding protein 1, DNA topoisomerase II-binding protein 1, KIAA0259DNA topoisomerase 2-binding protein 1, TOP2BP1, TopBP1, topoisomerase (DNA) II binding protein 1
Host Species Rabbit
Immunogen Maps to a region between residue 1400 and the C-terminus (residue 1435) of human Topoisomerase (DNA) II Binding Protein 1 using the numbering given in entry NP_008958.1 (GeneID 11073).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11073
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.