Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Topoisomerase I Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB130482AFC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB130482AFC 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB130482AFC Supplier Novus Biologicals Supplier No. NBP130482AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Topoisomerase I Polyclonal specifically detects Topoisomerase I in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Topoisomerase I
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DNA topoisomerase 1, DNA topoisomerase I, EC 5.99.1.2, TOPI, topoisomerase (DNA) I, type I DNA topoisomerase
Gene Symbols TOP1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 715 and 765 of human DNA topoisomerase I using the numbering given in entry NP_003277.1 (GeneID 7150).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 7150
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.