Learn More
Description
Specifications
Specifications
| Antigen | Topoisomerase I |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS with No Preservative |
| Gene Alias | DNA topoisomerase 1, DNA topoisomerase I, EC 5.99.1.2, TOPI, topoisomerase (DNA) I, type I DNA topoisomerase |
| Gene Symbols | TOP1 |
| Host Species | Rabbit |
| Immunogen | The immunogen for this product maps to a region between residue 715 and 765 of human DNA topoisomerase I using the numbering given in entry NP_003277.1 (GeneID 7150). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
