Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOPORS Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | TOPORS |
---|---|
Applications | Immunofluorescence, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2432
![]() |
Bethyl Laboratories, Inc
IHC00629T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2431
![]() |
Bethyl Laboratories, Inc
IHC00629 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Functions as an E3 ubiquitin-protein ligase and as an E3 SUMO1-protein ligase. Probable tumor suppressor involved in cell growth, cell proliferation and apoptosis that regulates p53/TP53 stability through ubiquitin-dependent degradation. May regulate chromatin modification through sumoylation of several chromatin modification-associated proteins. May be involved in DNA damage-induced cell death through IKBKE sumoylation.Specifications
TOPORS | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
LUN, p53-binding protein 3, P53BP3, retinitis pigmentosa 31 (autosomal domit), RP31, SUMO1-protein E3 ligase Topors, topoisomerase I binding, arginine/serine-rich, topoisomerase I binding, arginine/serine-rich, E3 ubiquitin protein ligase, Topoisomerase I-binding arginine/serine-rich protein, topoisomerase I-binding RING finger protein, TP53BPL, tumor suppressor p53-binding protein 3 | |
TOPORS | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q9NS56 | |
10210 | |
Between 995 and 1045 | |
Primary | |
4° C | |
TOPORS |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title