Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOR/mTOR Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB100240AFX Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100240AFX 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100240AFX Supplier Novus Biologicals Supplier No. NB100240AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TOR/mTOR Polyclonal specifically detects TOR/mTOR in Human, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen TOR/mTOR
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.11.1, FK506 binding protein 12-rapamycin associated protein 1, FK506 binding protein 12-rapamycin associated protein 2, FK506-binding protein 12-rapamycin complex-associated protein 1, FKBP12-rapamycin complex-associated protein, FLJ44809, FRAP, FRAP1FKBP12-rapamycin complex-associated protein 1, FRAP2, Mammalian target of rapamycin, Mechanistic target of rapamycin, mechanistic target of rapamycin (serine/threonine kinase), mTOR, RAFT1, rapamycin and FKBP12 target 1, rapamycin associated protein FRAP2, Rapamycin target protein 1, RAPT1FKBP-rapamycin associated protein, serine/threonine-protein kinase mTOR
Gene Symbols MTOR
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human Mammalian Target of Rapamycin using the numbering given in entry NP_004949.1 (GeneID 2475).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cell Cycle and Replication, DNA Repair, Hypoxia, Macroautophagy, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 2475
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.