Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOR1AIP2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB321725 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB321725 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB321725 Supplier Novus Biologicals Supplier No. NBP298140MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TOR1AIP2 Polyclonal antibody specifically detects TOR1AIP2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen TOR1AIP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias FLJ77012, IFRG15, interferon responsive gene 15, LULL1MGC120077, lumenal domain like LAP1, Lumenal domain-like LAP1, MGC120074, MGC120075, MGC120076, MGC126581, MGC138430, NET9, RP11-12M5.5, torsin A interacting protein 2, torsin-1A-interacting protein 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TOR1AIP2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163590
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.