Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOR3A Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 106471AP150UL

Description
TOR3A (Torsin Family 3 Member A) is a Protein Coding gene. Diseases associated with TOR3A include Apperceptive Agnosia and Speech And Communication Disorders. Gene Ontology (GO) annotations related to this gene include ATPase activity. An important paralog of this gene is TOR1A.Specifications
TOR3A | |
Polyclonal | |
Unconjugated | |
TOR3A | |
ADIR, ADIR2, TOR3A, Torsin 3A, Torsin family 3 member A, torsin family 3, member A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
30935, 64222 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9ER38, Q9H497 | |
TOR3A | |
TOR3A Fusion Protein Ag1012 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction