Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOR3A Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | TOR3A |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-6477
![]() ![]() |
Proteintech Group Inc
106471AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
TOR3A (Torsin Family 3 Member A) is a Protein Coding gene. Diseases associated with TOR3A include Apperceptive Agnosia and Speech And Communication Disorders. Gene Ontology (GO) annotations related to this gene include ATPase activity. An important paralog of this gene is TOR1A.Specifications
TOR3A | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q9ER38, Q9H497 | |
30935, 64222 | |
TOR3A Fusion Protein Ag1012 | |
Primary | |
-20°C | |
TOR3A |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ADIR, ADIR2, TOR3A, Torsin 3A, Torsin family 3 member A, torsin family 3, member A | |
TOR3A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title