Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TORC2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP222356 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222356 0.1 mL
NBP222356SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP222356 Supplier Novus Biologicals Supplier No. NBP222356
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TORC2 Polyclonal specifically detects TORC2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TORC2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1.0 - 2.0 ug/ml, Simple Western 1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:20 - 1:100, Immunohistochemistry-Paraffin 1:100
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q53ET0, Q3U182
Gene Alias CREB regulated transcription coactivator 2, CREB-regulated transcription coactivator 2, TORC-2, TORC2transducer of regulated cAMP response element-binding protein (CREB) 2, Transducer of CREB protein 2, Transducer of regulated cAMP response element-binding protein 2
Gene Symbols CRTC2
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human Torc2 protein (between residues 600-693) [UniProt Q53ET0]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Lipid and Metabolism, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 200186
Test Specificity Human and mouse. Predicted to react in rat and cow based on sequence homology.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.