Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TORC2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB337516 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337516 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337516 Supplier Novus Biologicals Supplier No. NBP222356MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TORC2 Polyclonal antibody specifically detects TORC2 in Human, Mouse samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TORC2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CREB regulated transcription coactivator 2, CREB-regulated transcription coactivator 2, TORC-2, TORC2transducer of regulated cAMP response element-binding protein (CREB) 2, Transducer of CREB protein 2, Transducer of regulated cAMP response element-binding protein 2
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human Torc2 protein (between residues 600-693) [UniProt Q53ET0]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Lipid and Metabolism, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 200186
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.