Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TORC3/CRTC3 Antibody (10G3), Novus Biologicals™
SDP

Catalog No. NB297688 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297688 50 μL
NB297689 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297688 Supplier Novus Biologicals Supplier No. NBP32664450UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TORC3/CRTC3 Monoclonal antibody specifically detects TORC3/CRTC3 in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen TORC3/CRTC3
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 10G3
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias CREB regulated transcription coactivator 3, FLJ21868, TORC-3, TORC3CREB-regulated transcription coactivator 3, Transducer of CREB protein 3, transducer of regulated cAMP response element-binding protein (CREB) 3, Transducer of regulated cAMP response element-binding protein 3, transducer of regulated CREB protein 3
Host Species Rabbit
Immunogen A synthesized peptide derived from Human TORC3/CRTC3 [UniProt Q6UUV7]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64784
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.