Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TORC3/CRTC3 Antibody (10G3), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | TORC3/CRTC3 |
---|---|
Clone | 10G3 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
TORC3/CRTC3 Monoclonal antibody specifically detects TORC3/CRTC3 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
TORC3/CRTC3 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
64784 | |
IgG | |
Affinity purified |
10G3 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
CREB regulated transcription coactivator 3, FLJ21868, TORC-3, TORC3CREB-regulated transcription coactivator 3, Transducer of CREB protein 3, transducer of regulated cAMP response element-binding protein (CREB) 3, Transducer of regulated cAMP response element-binding protein 3, transducer of regulated CREB protein 3 | |
A synthesized peptide derived from Human TORC3/CRTC3 [UniProt Q6UUV7] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title