Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Torin 1
GREENER_CHOICE

Catalog No. p-4817449
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective mTOR inhibitor

Torin 1 is a potent and selective ATP-competitive mTOR inhibitor (IC50 = 2 - 10 nM for mTORC1 and mTORC2). Torin 1 displays 200-fold selectivity for mTOR over DNA-PK, ATM and hVps34. In HeLa cells, Torin 1 induces autophagy. Torin 1 impairs cell growth and proliferation by suppression of the rapamycin-resistant functions of mTORC1. In human monocytes and myeloid dendritic cells, Torin 1 prevents decreases the anti-inflammatory potency of glucocorticoids. In the human endocrine cell line BON, Torin 1 increases neurotensin secretion and gene expression through MEK/ERK/c-Jun pathway activation.

Chemical Identifiers

CAS 1222998-36-8
Molecular Formula C35H28F3N5O2

Specifications

Quantity 10 mg
Formula Weight 607.62
Purity >98%
Molecular Weight (g/mol) 607.64
Chemical Name or Material Torin 1
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.