Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Torsin 2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP160002
Description
Torsin 2A Polyclonal specifically detects Torsin 2A in Human samples. It is validated for Western Blot.Specifications
| Torsin 2A | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FLJ14771, MGC99558, pro salusin, prosalusin, TORP1torsin-2A, Torsin family 2 member A, torsin family 2, member A, Torsin-2A, Torsin-related protein 1 | |
| Rabbit | |
| 36 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q5JU69 | |
| TOR2A | |
| Synthetic peptides corresponding to TOR2A(torsin family 2, member A) The peptide sequence was selected from the N terminal of TOR2A. Peptide sequence GLECDLAQHLAGQHLAKALVVKALKAFVRDPAPTKPLVLSLHGWTGTGKS. | |
| Protein A purified | |
| RUO | |
| 27433 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction