Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Torsin 2A Antibody, Novus Biologicals™
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Polyclonal Antibody
$159.00 - $487.50
Specifications
| Antigen | Torsin 2A | 
|---|---|
| Applications | Western Blot | 
| Classification | Polyclonal | 
| Conjugate | Unconjugated | 
| Form | Purified | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
                
                
                    
                        NBP16000220
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        Novus Biologicals
                         NBP16000220UL  | 
                
            
            
            
            
            
                
                    20 μL | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $159.00
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
                
                
                    
                        NBP160002
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        Novus Biologicals
                         NBP160002  | 
                
            
            
            
            
            
                
                    100 μL | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $487.50
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
Description
Torsin 2A Polyclonal specifically detects Torsin 2A in Human samples. It is validated for Western Blot.Specifications
| Torsin 2A | |
| Polyclonal | |
| Purified | |
| RUO | |
| FLJ14771, MGC99558, pro salusin, prosalusin, TORP1torsin-2A, Torsin family 2 member A, torsin family 2, member A, Torsin-2A, Torsin-related protein 1 | |
| TOR2A | |
| IgG | |
| Protein A purified | 
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Q5JU69 | |
| 27433 | |
| Synthetic peptides corresponding to TOR2A(torsin family 2, member A) The peptide sequence was selected from the N terminal of TOR2A. Peptide sequence GLECDLAQHLAGQHLAKALVVKALKAFVRDPAPTKPLVLSLHGWTGTGKS. | |
| Primary | |
| 36 kDa | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title