Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Total Bilirubin Reagents
Click to view available options
Quantity:
1 x 120 mL
1 x 1 L
Description
Includes
Total bilirubin reagent: Sulfanilic acid 16mM, hydrochloric acid 164mM, dimethyl sulfoxide 4.4M, surfactant; Sodium nitrite reagent: Sodium nitrite 0.6M
- Method: Walters and Gerarde, Endpoint
- Format: Liquid (ready to use)
- Wavelength: 550nm
- Linearity: 20mg/dL
- Expected Values: 0.2 to 1.2mg/dL
For the quantitative determination of total bilirubin in serum.
Specifications
Specifications
Certifications/Compliance | CE Marked |
Type | Total Bilirubin Reagent Kit |
Formulation | Total Bilirubin reagent: Sulfanilic Acid 16mM, Hydrochloric Acid 164mM, Dimethyl Sulfoxide 4.4M, Surfactants, Sodium Nitrite 0.6M |
For Use With (Application) | Quantitative determination of total Bilirubin in Serum, in-vitro diagnostic use only |
Quantity | 1 x 1 L |
Wavelength | 550 nm |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction