Learn More
Ansell TouchNTuff™ 69-318 Lightweight Latex Gloves
Catalog No. 19050060B Shop All Ansell Products
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Lightweight latex glove that offers textured fingers and tactile sensitivity for easy handling of small objects and delicate instruments.
  • Manufactured from natural rubber latex
  • Thin, lightweight design
  • Powder free
  • Soft, elastic material provides comfort
  • Primiarily used in life science, chemical and food industries
  • Complies with FDA food handling requirements
TRUSTED_SUSTAINABILITY
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.