Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ XtraTuff™ Rough Textured Oil-Resistant PVC Gloves

Catalog No. 19023123B Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Medium
Large
X-Large
Length (English):
10 in.
12 in.
This item is not returnable. View return policy
This item is not returnable. View return policy

Oil-resistant gloves stay soft and strong, even after exposure to machine oils. Ideal for applications that require protection against cuts and liquids, with a rough textured palm for enhanced grip capabilities.

  • Oil-resistant gloves remain soft and strong, even after exposure to machine oils
  • Textured palm enhances wet and dry grip capabilities
  • Seamless liner is treated to resist germs and odors
  • Gauntlet cuff extends protection up to the forearm
  • Ideal for applications that require protection against cuts and liquids

Specifications

Certifications/Compliance FDA Part 177, Subpart C
Coating Material PVC
Color Blue
Exterior Finish Fully Textured
Hand Specific
Latex Free Yes
Material PVC
Product Type Dip-coated Gloves
Size Medium
EN388 Standard 4121
For Use With (Application) Aquatics/Marine Research, Handling Oily Parts
Glove Cuff Style Gauntlet
Liner Material Cotton
Series 58-8656
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.