Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOX2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP258503 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP258503 100 μL
NB393421 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP258503 Supplier Novus Biologicals Supplier No. NBP258503
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TOX2 Polyclonal specifically detects TOX2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TOX2
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias C20orf100, chromosome 20 open reading frame 100, dJ495O3.1, GCX1, GCX-1dJ1108D11.2, granulosa cell HMG box 1, Granulosa cell HMG box protein 1, granulosa cell HMG-box protein 1, MGC15880, TOX high mobility group box family member 2
Gene Symbols TOX2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:HEASYHSLCHGLTPNGLLPAYSYQAMDLPAIMVSNMLAQDSHLLSGQLPTIQEMVHSEVAAYDSGR
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84969
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.