Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOX3 Antibody (TOX3/1123), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2451650.1MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
TOX3 Monoclonal specifically detects TOX3 in Human, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array, Immunofluorescence.Specifications
TOX3 | |
Monoclonal | |
0.2mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
O15405 | |
TOX3 | |
Recombinant fragment (139 Amino acid residues around aa 200-400) of human TOX3 protein | |
Protein A or G purified | |
RUO | |
27324 | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
TOX3/1123 | |
Unconjugated | |
1.0mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CAG trinucleotide repeat-containing gene F9 protein, CAGF9trinucleotide repeat containing 9, TNRC9, TOX high mobility group box family member 3, Trinucleotide repeat-containing gene 9 protein | |
Mouse | |
63 kDa | |
0.1 mg | |
Primary | |
It recognizes a 63kDa protein, which is identified as TOX3. It contains a high mobility group (HMG)-box, which regulates Ca2+-dependent transcription in neurons through interaction with the cAMP-response-element-binding protein (CREB). TOX3 appears to be associated with breast cancer susceptibility and is expressed downstream of a cytoprotective cascade together with CITED1, a transcriptional regulator that does not bind directly to DNA. TOX3 is predominantly expressed in the brain and forms homodimers. TOX3 overexpression protects neuronal cells from cell death caused by endoplasmic reticulum stress or BAX overexpression through the induction of anti-apoptotic transcripts and repression of pro-apoptotic transcripts. | |
Store at 4C. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction