Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOX4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP180328 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP180328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP180328 Supplier Novus Biologicals Supplier No. NBP180328
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TOX4 Polyclonal specifically detects TOX4 in Human samples. It is validated for Western Blot.

Specifications

Antigen TOX4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_055643
Gene Alias C14orf92, chromosome 14 open reading frame 92, Epidermal Langerhans cell protein LCP1, KIAA0737MIG7, LCP1, migration-inducing protein 7, TOX high mobility group box family member 4
Gene Symbols TOX4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal of human KIAA0737. Peptide sequence MEFPGGNDNYLTITGPSHPFLSGAETFHTPSLGDEEFEIPPISLDSDPSL.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9878
Test Specificity Expected identity based on immunogen sequence: Zebrafish: 100%; Rat: 100%; Sumatran orangutan: 100%; Green puffer: 100%; Mouse: 100%; Bovine: 100%; Western clawed frog: 100%; Human: 100%; Xenopus: 92%; Florida lancelet: 91%; Chicken: 91%; Pig: 91%; Black-legged tick: 90%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.