Learn More
Toxoplasma Gondii P30 Antibody (P30/3), Allophycocyanin/Cy7, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Toxoplasma Gondii P30 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | P30/3 |
| Conjugate | APC-Cyanine7 |
| Dilution | Flow Cytometry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | p30, SAG1, SAG1 protein |
| Host Species | Mouse |
| Immunogen | Purified T. gondii P30 antigen |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.