Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ TP 064

Click to view available options
Quantity:
10 mg
50 mg
Description
TP 064 is a potent and selective PRMT4 inhibitor; inhibits methylation of H3 (1-25) and MED12 (IC50 values are <10 and 43 nM, respectively). Exhibits >100-fold selectivity over other histone methyltransferases and non-epigenetic targets. Inhibits proliferation of certain multiple myeloma cell lines, arresting cells in G1 phase. TP 064 can be used in vivo. TP 064 inhibits the monocyte/macrophage-driven inflammation in ex vivo.
To request the negative control for TP 064, please fill out the TP 064N request form on the SGC website..
Specifications
Specifications
| Chemical Name or Material | TP 064 |
| Synonym | N-Methyl-N-((2-(1-(2-(methylamino)ethyl)piperidin-4-yl)pyridin-4-yl)methyl)-3-phenoxybenzamide |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in 1eq. HCl and to 100mM in DMSO |
| Molecular Formula | C28H34N4O2 |
| Formula Weight | 458.6 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction