Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TP53INP2 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP171704B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP171704B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP171704B Supplier Novus Biologicals Supplier No. NBP171704B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TP53INP2 Polyclonal specifically detects TP53INP2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TP53INP2
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias C20orf110, chromosome 20 open reading frame 110, diabetes and obesity regulated, dJ1181N3.1, DKFZp434B2411, DKFZp434O0827, DOR, FLJ21759, FLJ23500, p53-inducible protein U, PINHPIG-U, tumor protein p53 inducible nuclear protein 2, tumor protein p53-inducible nuclear protein 2
Gene Symbols TP53INP2
Host Species Rabbit
Immunogen A genomic peptide made to a C-terminal region of the human TP53INP2 protein (within residues 100-220). [Swiss-Prot Q8IXH6]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 58476
Test Specificity Human and mouse. Immunogen has 81% identity to rat.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.