Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TP53TG5 Antibody (Z38P1E4*C7), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP250338V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250338V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250338V Supplier Novus Biologicals Supplier No. NBP250338V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TP53TG5 Monoclonal specifically detects TP53TG5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TP53TG5
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Z38P1E4*C7
Conjugate DyLight 405
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C20orf10, chromosome 20 open reading frame 10, CLG01, dJ453C12.5, TP53 target 5, TP53-inducible gene 5 protein, TP53-target gene 5 protein
Gene Symbols TP53TG5
Host Species Mouse
Immunogen Peptide Sequence - WRNSRVYK
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27296
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.