Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TP73-AS1 Antibody (OTI2B9), Novus Biologicals™
SDP

Catalog No. NBP201283 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP201283 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP201283 Supplier Novus Biologicals Supplier No. NBP201283
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TP73-AS1 Monoclonal specifically detects TP73-AS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TP73-AS1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2B9
Concentration 0.65 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150
Gene Alias KIAA0495, MGC138189, PDAM
Gene Symbols TP73-AS1
Host Species Mouse
Immunogen Full length human recombinant protein of human KIAA0495(NP_997198)
Molecular Weight of Antigen 20.4 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57212
Target Species Human, Monkey
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.