Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPD52 Antibody, Novus Biologicals™
SDP

Catalog No. NBP106090 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106090 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106090 Supplier Novus Biologicals Supplier No. NBP106090
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

TPD52 Polyclonal specifically detects TPD52 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen TPD52
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001020423.1
Gene Alias D52, hD52, N8L, PC-1, PrLZ, prostate and colon associated protein, prostate leucine zipper, Protein N8, tumor protein D52
Gene Symbols TPD52
Host Species Goat
Immunogen Peptide with sequence C-TEPLPEKTQES corresponding to C-Terminus according to NP_001020423.1 NP_001020424.1, NP_005070.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7163
Test Specificity This antibody is expected to recognize all three reported isoforms (NP_001020423.1, NP_001020424.1and NP_005070.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.