Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPMT Antibody (2E7), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP197939AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197939AI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197939AI Supplier Novus Biologicals Supplier No. NBP197939AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TPMT Monoclonal specifically detects TPMT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TPMT
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 20000000
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.1.1.67, S-adenosyl-L-methionine:thiopurine S-methyltransferase, Thiopurine methyltransferase, thiopurine S-methyltransferase
Gene Symbols TPMT
Host Species Mouse
Immunogen Recombinant human TPMT (1-245aa) purified from E. coli
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7172
Test Specificity Anti-human TPMT mAb, clone 2E7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human TPMT protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.