Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPPP2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB080604 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB080604 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB080604 Supplier Novus Biologicals Supplier No. NBP297003AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TPPP2 Polyclonal specifically detects TPPP2 in Human samples. It is validated for ELISA.

Specifications

Antigen TPPP2
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias C14orf8, chromosome 14 open reading frame 8, P18, p25beta, Protein p25-beta, TPPP/p18, tubulin polymerization-promoting protein family member 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant TPPP2 (AAH38970.1; Met1-Lys170).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122664
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.