Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPRKB Antibody (OTI3H3), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP27474A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27474A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27474A32 Supplier Novus Biologicals Supplier No. NBP274574AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TPRKB Monoclonal specifically detects TPRKB in Human, Mouse, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TPRKB
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3H3
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CGI-121, PRPK (p53-related protein kinase)-binding protein, PRPK-binding protein, TP53RK binding protein, TP53RK-binding protein
Gene Symbols TPRKB
Host Species Mouse
Immunogen Full length recombinant protein of human TPRKB (NP_057142) produced in E.coli.
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51002
Target Species Human, Mouse, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.