Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptase alpha/TPS1 Rabbit anti-Human, Mouse, Rat, Clone: SC68-07, Novus Biologicals™
SDP

Catalog No. NBP267608 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267608 Supplier Novus Biologicals Supplier No. NBP267608
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Tryptase alpha/TPS1 Monoclonal specifically detects Tryptase alpha/TPS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen TPSAB1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SC68-07
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias EC 3.4.21, EC 3.4.21.59, mast cell alpha II tryptase, mast cell beta I tryptase, MGC104064, TPS1tryptase alpha II, TPS2, TPSB1, tryptase alpha/beta 1, tryptase alpha/beta-1, Tryptase alpha-1, tryptase beta 1, tryptase beta I, tryptase beta-1, Tryptase I, tryptase-1, tryptase-I, tryptase-III
Gene Symbols TPSAB1
Host Species Rabbit
Immunogen Recombinant protein within Human Tryptase alpha/TPS1 aa 10-275/ 275. (SwissProt: Q15661 Human; SwissProt: Q02844 Mouse; SwissProt: P27435 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Immunology, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7177
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.