Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPX2 Antibody (TPX2-01), HRP, Novus Biologicals™
SDP

Catalog No. NBP267265H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267265H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267265H Supplier Novus Biologicals Supplier No. NBP267265H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TPX2 Monoclonal specifically detects TPX2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen TPX2
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone TPX2-01
Concentration 1mg/mL
Conjugate HRP
Dilution Western Blot, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS with No Preservative
Gene Alias C20orf1HCTP4, C20orf2, chromosome 20 open reading frame 1, Differentially expressed in cancerous and non-cancerous lung cells 2, differentially expressed in lung cells, DIL2, DIL-2FLS353, HCA519, Hepatocellular carcinoma-associated antigen 519, P100, p100GD:C20orf1, preferentially expressed in colorectal cancer, Protein fls353, REPP86, restricted expression proliferation associated protein 100, Restricted expression proliferation-associated protein 100, targeting protein for Xklp2, TPX2, microtubule-associated protein homolog, TPX2, microtubule-associated, homolog (Xenopus laevis)
Gene Symbols TPX2
Host Species Mouse
Immunogen Human TPX2
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, DNA Repair, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 22974
Test Specificity The mouse monoclonal antibody TPX2-01 recognizes the epitope EPFVPKKEKKS (aa 636-646 in human) of TPX2, a microtubule-associated intracellular critical regulator of mitosis, which is overexpressed in many types of tumors and is a marker of worse prognosis in various cancers.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.