Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TR alpha/NR1A1/Thyroid Hormone Receptor alpha Antibody (1330), Novus Biologicals™
SDP

Catalog No. NB10061087 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10061087 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10061087 Supplier Novus Biologicals Supplier No. NB10061087
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TR alpha/NR1A1/Thyroid Hormone Receptor alpha Monoclonal specifically detects TR alpha/NR1A1/Thyroid Hormone Receptor alpha in Human, Rat samples. It is validated for Western Blot.

Specifications

Antigen TR alpha/NR1A1/Thyroid Hormone Receptor alpha
Applications Western Blot
Classification Monoclonal
Clone 1330
Conjugate Unconjugated
Dilution Western Blot 1:1000
Gene Accession No. P10827
Gene Alias AR7, c-ERBA-1, c-erbA-alpha, EAR7, EAR-7, EAR-7.1/EAR-7.2, ERBA, ERBA1thyroid hormone receptor, alpha (avian erythroblastic leukemia viral (v-erb-a)oncogene homolog), ERBA-related 7, MGC000261, MGC43240, NR1A 1, NR1A1THRA3, Nuclear receptor subfamily 1 group A member 1, THRA1ERB-T-1, THRA2, thyroid hormone receptor alpha, thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a)oncogene homolog, avian), triiodothyronine receptor, V-erbA-related protein 7
Gene Symbols THRA
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acid residues from the N-terminal region conjugated to KLH. Accession # P10827
Molecular Weight of Antigen 58 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7067
Test Specificity Specific for the ∼58k TR-alpha 2 protein.
Target Species Human, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.