Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TR beta 1/NR1A2/Thyroid Hormone Receptor beta Antibody, Novus Biologicals™
SDP

Catalog No. NB10093535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093535 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093535 Supplier Novus Biologicals Supplier No. NB10093535
Add to Cart
Add to Cart

Goat Polyclonal Antibody

TR beta 1/NR1A2/Thyroid Hormone Receptor beta Polyclonal specifically detects TR beta 1/NR1A2/Thyroid Hormone Receptor beta in Zebrafish samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen TR beta 1/NR1A2/Thyroid Hormone Receptor beta
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 - 5 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_571415.1
Gene Alias avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, c-erbA-2, c-erbA-beta, ERBA2MGC126109, ERBA-BETA, GRTH, MGC126110, NR1A2thyroid hormone receptor, beta (avian erythroblastic leukemia viral (v-erb-a)oncogene homolog 2), Nuclear receptor subfamily 1 group A member 2, oncogene ERBA2, PRTH, THR1pituitary resistance to thyroid hormone, THRB1, THRB2, thyroid hormone receptor beta, thyroid hormone receptor beta 1, thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a)oncogene homolog 2, avian)
Gene Symbols THRB
Host Species Goat
Immunogen Peptide with sequence C-DIGSAPIVNAPEGNKVD corresponding to internal region according to Uniprot Zebrafish NP_571415.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7068
Test Specificity This antibody is not expected to cross-react to Thra.
Target Species Zebrafish
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.