Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TR2/NR2C1 Antibody (OTI4C7), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP27476A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27476A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27476A32 Supplier Novus Biologicals Supplier No. NBP274576AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TR2/NR2C1 Monoclonal specifically detects TR2/NR2C1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen TR2/NR2C1
Applications Western Blot
Classification Monoclonal
Clone OTI4C7
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias nuclear receptor subfamily 2 group C member 1, nuclear receptor subfamily 2, group C isoform, nuclear receptor subfamily 2, group C, member 1, Orphan nuclear receptor TR2, Testicular receptor 2, TR2, TR2 nuclear hormone receptor, TR2-11
Gene Symbols NR2C1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-237 of human TR2/NR2C1 (NP_001027458) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7181.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.